General description
Vivapure spin columns employ an innovative and powerful ion exchange membrane technology for purification of proteins. Vivapure purification protocols allow the isolation of ultrapure proteins with high yield in less than 30 minutes.
The columns are available with different ion exchange resins:
• Type C: carboxylic acid (weak acidic cation exchanger)
• Type D: diethylamine (weak basic cation exchanger)
• Type Q: quarternary ammonium (strong basic cation exchanger)
• Type S: sulphonic acid (strong acidic cation exchanger)
Note: Comes with comphrensive instruction handbook and laminated protocol card.