Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
Z630675
Sigma
Binder small incubator B28 series
with natural convection
zoom
1 of 1
Expand/Collapse All
Price and Availability
Properties
AC input
230 V
material
glass inner door
feature
CE compliant
with natural convection
availability
available only in EU
limit
30-70 °C temperature
L × W × H
425 mm × 580 mm × 402 mm
inner chamber volume
28 L
weight
16 kg
Related Categories
...
Binder
>
Incubators
...
Microbiology Equipment
>
Incubators
...
Heating
>
Ovens
Scroll Up
Scroll Down
Related Information
MSDS
Certificate of Analysis
More Information
Links
Similar Products
Related Categories
Page Options
Printer Friendly View
Ask A Scientist
Email Page
Last 5 Products Viewed
Z630675 (Sigma)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.