General description
Particularly high requirements are made on temperature accuracies during hybridisation. The BFD series ovens are efficient, whilst including a broad temperature range that they are also used without limitation as incubators and heating chambers. The high quality outfitting and the expedient inner chamber volume underlines their versatility. Perfect ease of cleaning and simple operation complete the reliability of the unit.
• AP-Line Preheating Chamber Technology
• 5 °C above room temperature to 99.9 °C
• Powerful air turbine
• Digital temperature setting with tenth of a degree accuracy
• Timer 0 - 24 hour
• Removable, electrical rotation unit for up to 12 hybridisation bottles with automatic switch-off
• Chromed rack
• Removable drip pan
• Inner glass door with smooth inner side
• Temperature safety device CI. 3.1
• Adjustable ventilation via rear exhaust duct diam. 50 mm with ventilation flap and front ventilation slide