General description
Chamber particularly suitable for hybridisation in bags, incubation of flasks whilst shaking, tempering of wash bags and in-situ hybridisation. The shaking frequency can be set digitally and can accommodate different bottle holders.
• AP-Line Preheating Chamber Technology
• 5 °C above room temperature to 99.9 °C
• Powerful air turbine
• Digital temperature setting with tenth of degree accuracy
• Timer 0 -99.59 hour
• Removable, electrical rotation unit for up to 12 hybridisation bottles with automatic switch-off
• Chromed rack
• Removable drip pan
• Inner glass door with smooth inner side
• Temperature safety device CI. 3.1
• Adjustable ventilation via rear exhaust duct diam. 50mm with ventilation flap and front ventilation slide
RS232 printer and communication interface for standard software APT-COM DataControlSystem