General description
Consists of a clear plexiglas block with 12x extra strong permanent magnets for a fast magnetic separation with 1.5 mL and 2.0 mL tubes. With 48 tube positions including 24 positions for magnetic separation (12x for 1.5 mL and 12x for 2.0 mL tubes). For 1.5 mL (conical) and 2.0 mL (conical recommended) tubes. Transparent material allows for constant visual control of the separation. For fast separation (< 2 min).
Legal Information
BILATEST is a registered trademark of Bilatec AG.