General description
Ohaus Voyager Pro balances offer features including:
• Autocal feature - automatically re-calibrates if temperature change sufficient to effect accuracy
• Ergonomic levelling system
• Advanced applications function eliminates the need for time consuming manual time calculations
• Simple operation - one function per key
• Removable draft shield doors
• weighing units include g, kg, oz, lb, pound, ounce, ozt, dwt
• RS232 interface port
• Sealed front panel and moulded spill ring to protect the balance during use
• Lockswitch to allow balance to be locked into a specific configuration, reducing operator error
Legal Information
Ohaus is a registered trademark of Ohaus Corp.