Features and Benefits
• Highest performance
• Multiple weighing units
• Environmental settings
• Easy to set up and use
• Advanced AutoCal calibration
• Four methods of density determination
• RRS232 interface with full GLP/GMP Protocol
• Protective in-use cover
• Up-front level indicator
General description
Ohaus Discovery series of semi-micro and analytical balances combine unmatched weighing performance with Ohaus' innovative SmarText software, making it extremely reliable and easy-to-use. The Discovery features durable glass and steel construction and together with advanced AutoCal internal calibration makes it the best performing balance in its class.
The four Discovery models have capacities upto 310g by 0.1mg with a FineRange model that offers 10 microgram readability are perfect for university research, pharmaceutical research and drug discovery, and general lab applications.