Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
Z667161
Sigma
Greiner PS inoculating needle
zoom
1 of 1
Expand/Collapse All
Price and Availability
Properties
material
yellow polystyrene
sterility
sterile
mfr. no.
Greiner 731180
availability
available only in EU
L
200 mm
Related Categories
...
Cell Culture Supplies
>
Inoculation
...
Culture
>
Inoculation
...
Microbiology Equipment
>
Inoculation
Scroll Up
Scroll Down
Related Information
Needle Gauge Guide
MSDS
Certificate of Analysis
More Information
Links
Related Categories
Page Options
Printer Friendly View
Ask A Scientist
Email Page
Last 5 Products Viewed
Z667161 (Sigma)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.