General description
Ohaus Adventurer Pro balance offer features including:
• Brilliant backlit high resolution dot matrix display
• RS232 port for easy communication with computer or printer
• GLP compliant with user selectable outputs
• AC adapter or 4 "AA" batteries (not supplied)
• sealed front panel and moulded spill ring to protect the balance during use
• includes an integral shipping lock to protect the weighing mechanism during transportation
• lockswitch to allow balance to be locked into a specific configuration, reducing operator error
• Industry leading warranty and support
Legal Information
Adventurer is a trademark of Ohaus Corp.
Ohaus is a registered trademark of Ohaus Corp.