General description
Proteins interacting with diverse ligands - proteins, peptides or DNA - are the basic principles underlying many biological processes, such as antigen-antibody binding, signal transduction or receptor binding. The technique of oligopeptide synthesis on a cellulose membrane and the subsequent binding assays allow the investigation of protein interactions. A particular advantage of these peptide arrays (SPOT - technology) is the high number of oligopeptide probes that can be tested in parallel. Detailed protocols for peptide synthesis, and the analysis of protein-protein, protein-DNA interactions as well as epitope mapping are presented.