General description
DNA Microarrays are an indispensable tool in molecular biology, which allows researchers to gain insight into the intricate mechanisms of gene expression. Common to all array-based technologies is the necessity to analyze digital images of the scanned DNA array. This book emphasizes aspects of both theory and application. It offers the basics of machine vision and image analysis for biologists, "how-to" approach to microarray image analysis, and practical tips for both the novice in the field and the advanced user.
Table of Contents
Chapter 1. Microarray and Image Analysis: Introduction
Chapter 2. Introduction to Image Analysis Chapter 3. Advantages of Laser Confocal Microarray Scanning Chapter 4. Considerations for a Quality Microarray Scanner Chapter 5. Key Considerations for Accurate Microarray Scanning and Image Analysis Chapter 6. Microarray Image Processing and Quality Control Chapter 7. Microarray Data Normalization Chapter 8. Quantitative Comparison of Image Analysis Software Chapter 9. DNA Array Information Workflow and Data Management Chapter 10. BAC Microarrays Index
| ||||||||