General description
A revised and updated collection readily reproducible techniques for culturing human cells. This edition includes a wide range of human cell types relevant to human disease and new chapters on fibroblasts, Schwann cells, gastric and colonic epithelial cells, and parathyroid cells. The protocols follow the successful Methods in Molecular Medicine¿ series format, each offering step-by-step laboratory instructions, an introduction outlining the principle behind the technique, lists of the necessary equipment and reagents, and tips on troubleshooting and avoiding known pitfalls