General description
A collection of key cytogenetic and FISH techniques used by modern clinical laboratories in the genetic analysis of human malignancies. The book's practical advice and methods are suitable for use at every level of expertise, including fully established laboratories, but with a sympathetic bias towards anyone considering setting up a new cytogenetics service. Here the reader will find not only elementary tutorials on the fundamentals of human karyotypes and chromosome analysis, but also detailed discussions on how laboratories may optimally upgrade their repertoire of capabilities to include such newer complementary techniques as CGH, FISH, and M-FISH.
Table of Contents
1. Introduction
2. Cytogenetic Studies in Hematological Malignancies: an Overview 3. The Myeloid Disorders: Background 4. Cytogenetic Techniques for Myeloid Disorders 5. Acute Lymphoblastic Leukemia: Background 6. Conventional Cytogenetic Techniques in the Diagnosis of Childhood Acute Lymphoblastic Leukemia 7. Chromosome Preparations from Bone Marrow in Acute Lymphoblastic Leukemia 8. Lymphoid Disorders Other than Common Acute Lymphoblastic Leukemia: Background 9. Other Lymphoid Malignancies: Cytogenetic Techniques 10. Cytogenetic and Genetic Studies in Solid Tumors: Background 11. Human Solid Tumors: Cytogenetic Techniques 12. Analysis and Interpretation of Cytogenetic Findings in Malignancies 13. Cytogenetic Studies Using FISH: Background 14. FISH Techniques 15. FISH, CGH, and SKY in the Diagnosis of Childhood Acute Lymphoblastic Leukemia 16. Solving Problems in Multiplex FISH 17. Some Difficult Choices in Cytogenetics 18. Inroduction to the Analysis of the Human G-Banded Karyotype Index
| ||||||||