General description
TotalLab is a highly automated and flexible image analysis software package. It offers the complete solution for all your general image analysis requirements across a wide range of applications.
There are modules for:
• 1D electrophoresis gel analysis - Single click identification of lanes and bands, and accurate calculation of molecular weights from even the most distorted 1D images.
• Array analysis - Ideal for the quantitative analysis of dot and slot blots, microtitre plates and other basic arrays, creating grids of up to 1,536 cells.
• Colony counting - Accurate counts in seconds, adjustable parameters and editing tools if fine tuning is required.
• General image analysis using Toolbox - General quantitation on any image from a wide range of biological samples using the easy-to-use range of tools provided.
The analysis work-flow within the modules is the same throughout. The interface follows a wizard-style making it easy for you to navigate through the analysis, there are Help Panes at each stage and parameters for the analysis are easily accessible to assist with the refinement of the analysis.
System Requirements
Windows 2000, Windows XP
Processor: 1.4 GHz
Memory: minimum 256 MB, recommended 512 Mb
Free hard disk space: Recommended 5 Gb