General description
2D electrophoresis is a key technique within life science research to separate and analyse protein mixtures. The use of 2D electrophoresis has evolved dramatically both in terms of the number of scientists utilising the technique and how it is applied within their research. As a result new technology is continually being developed and implemented by Nonlinear to meet the growing needs of researchers. This is an entry level product for quick, simple interpretation of 2D gel image analysis
• Analysis wizard for simple setup of gel processing
• Unique algorithms to help eliminate major noise features from affecting spot detection and matching
• Spot detection that captures all spot material, with automatic splitting of overlapping spots
• Automatic warping to drive accurate spot matching
• Spot filters, manual warping and a suite of simple to use tools for edting analysis results
• Automatic generation of t-test and ANOVA results
• Sort and filter measurement to only display spots with interesting profiles
• 2D and 3D image