General description
This reference contains comprehensive information on more than 3,000 solvents. It serves as an integrated reference tool for decision making in formulation, purchasing, and the understanding of safety issues. Each entry contains information on: chemical composition, classification, properties, functions/applications, toxicology, flammability and reactivity ratings, storage, environmental impact for both trade name and generic solvents that are manufactured worldwide. A functional/application cross-reference contains more than 10,000 categories followed by an alphabetical listing of the trade name products and chemicals that are suited for use in these areas.