General description
Progenesis PG200 provides simple laboratory image analysis for single stain 2D gels using a routine, three step approach – Set-up, Process and Review. With this approach an analysis is set-up in minutes, processed automatically and reviewed using a comprehensive suite of display options to show spots of interest. Analysis workflows can be tailored to the specific requirements of your lab using an intuitive user interface and step by step approach. Key features of PG200 include
• Analysis Wizard to set-up entire an experiment for analysis
• Automated Matching with option for manual seed placement
• Background subtraction that is flexible and fully informative
• Normalisation methods to perform accurate intensity calibration and MW/pl determination on your 2D gels
• Editing tools to optimize analysis results
• SameSpots spot outline method, which offers a brand new approach for analysing aligned images
• Image views and measurements tables that highlight areas of interest and present results
• Workspaces that can be recalled to display custom window layouts for easy data review
System Requirements
Operating system: Windows 2000 and Windows XP
Processor: Minimum Pentium 4 2.0 GHz, recommended Pentium 4 3.0 GHz
Memory: Minimum 1 Gb, recommended 2 Gb
Free hard disk space: Recommended 5 Gb
Recommended graphics cards: Nvidia - GeForce 2, GeForce 3, GeForce 4, GeForce FX, Quadro 2, Quadro 4, Quadro FX
ATI.Diamond - Fire GL