General description
A revolutionary tagged primer design tool for expression cloning.
• High throughput primer design- Use Xpression Primer's sophisticated algorithm to design thousands of primers in a single search run. The program uses statistical optimization techniques to select the best primer pair for each sequence based on your selection criteria.
• Extensive Assay Support- Xpression Primer provides comprehensive support to high throughput cloning systems such as Gateway™, BD In-Fusion, epitope and TOPO™ tools. To work with other expression systems, add functional tags of your choice and design tagged primers.
• Sophisticated Sequencing Primer design- Xpression Primer designs optimal primers for multiples sequences in a single search run.
• Generate Precise transcripts- Design primers to generate sense or antisense transcripts for in vitro expression studies.
• Web Savvy- Xpression Primer searches Entrez and downloads batches of ORF sequences directly into the program.
• Lightning fast- Processes three hundred 10kb sequences in approximately 2 minutes.
Legal Information
TOPO is a registered trademark of Invitrogen Corp.