General description
A significantly revised and improved edition, this ediition provides the essential information required to understand and apply electrochemical processes. It offers a basic outline of the essential topics of theoretical and applied electrochemistry and serves as a key to recent and advanced developments in the field. This new edition features a new focus on experimental techniques, including a comprehensive chapter on physical methods for the investigation of electrode surfaces. New chapters deal with recent trends in electrochemistry, including nanoelectrochemistry, solid-state electrochemistry, conducting polymers, electrocatalysis, computer simulation in electrochemistry, and electrochemistry and the environment. Chapters included on batteries, fuel cells, supercapacitors, and other electrochemical devices and phenomena.