General description
Experts summarize the latest discoveries concerning the changes that occur in tumor cells as they develop resistance to anticancer drugs, and suggest new approaches to preventing and overcoming it. The authors review physiological resistance based upon tumor architecture, cellular resistance based on drug transport, epigenetic changes that neutralize or bypass drug cytotoxicity, and genetic changes that alter drug target molecules by decreasing or eliminating drug binding and efficacy. Highlights include new insights into resistance to antiangiogenic therapies, oncogenes and tumor suppressor genes in therapeutic resistance, cancer stem cells, and the development of more effective therapies. There are also new findings on gene amplification in drug resistance, and the molecular determinants of multidrug resistance.