Compatibility
Uses Finntip 250 Univ or 200 Ext pipet tips
General description
Convenience and comfort are designed into the Finnpipette Focus around the most important feature - extremely accurate and precise pipetting. The unique design specifically reduces user stress and optimizes pipetting performance so that you can set the volumes you need quickly and focus on your work.
• Unique snap-on handle cover in three different sizes (S,M,L)
• Fast volume adjustment
• Fine adjustment
• Soft-touch tip ejection
• Advanced innovative design
• Short tip cone
• Autoclavable
Legal Information
Finnpipette is a registered trademark of Thermo Electron Corp.