Compatibility
Uses Finntip 5 mL pipet tips
General description
The Finnpipette Focus Fixed Volume pipettes offer the same ergonomic advantages as the rest of the Finnpipette Focus range, including soft-touch tip ejection and the short precision control tip. Finnpipette Fixed Volume pipettes are provided with blue medium sized handles. The entire pipette including the tip cone and snap-on handle can be autoclaved in one piece
Legal Information
Finnpipette is a registered trademark of Thermo Electron Corp.