General description
Ohaus Scout Pro balances offer features including:
• weighing units g, kg, oz, lb, pound, ounce, ozt, dwt
• RS232 interface with integral cable
• two-button keypad and enhanced user interface speeds up use
• change weighing units with a touch of a button
• AC adapter or 4 "AA" batteries
• sealed front panel and moulded spill ring to protect the balance during use
• includes an integral shipping lock to protect the weighing mechanism during transportation
• lockswitch to allow balance to be locked into a specific configuration, reducing operator error
Legal Information
Scout is a registered trademark of Ohaus Corp.
Ohaus is a registered trademark of Ohaus Corp.