Compatibility
Compatibility
Biohit Electronic (Limited Draw) 5-100 μl
Biohit Variable 5-50 μl
Biohit Multi-Channel 5-50 μl
Capp Aero 50 μl
Eppendorf Research Pro 4860 5-100 μl
Eppendorf Titerman 4908 5-50 μl
Finnpipette Multi-Channel 5-50 μl
Rainin EDP-Series 100 (Limited Draw) 10-100 μl
Socorex/Wheaton 822 Series (Limited Draw) 5-100 μl
General description
Universal SoftFit™ Design to fit most 50 μl multi-channel pipettors
Precision-molded tips with the patented Art self-sealing barrier. Guaranteed to prevent carryover contamination during aspiration and delivery of samples. Art tips are ideal for use in genetic studies, forensics, general PCR work and radioisotope sampling. Art tips are available in numerous sizes and styles to meet all your liquid handling needs. All Art tips are certified RNase-, DNase-, DNA- and pyrogen-free.
Legal Information
SoftFit is a trademark of Molecular BioProducts, Inc.
ART is a registered trademark of Molecular BioProducts, Inc.