Compatibility
Compatibility
Biohit Proline-Electronic 10-100 μl
Eppendorf 4810 2-20 μl
Eppendorf Research/Reference 2-20 μl
Finnpipette 4500 Digital .5-10 μl
Hamilton SoftGrip Precision 25 μl
Mettler-Toledo VoluMate 2-20 μl
Nichriyo/Oxford BenchMate 2-20 μl
Pipetman P-20 2-20 μl
Rainin EDP 2 2.5-25 μl
Rainin EDP Plus 2.5-25 μl
Rainin Pipet Plus 2.5-20 μl
Rainin Pipetman 2.5-20 μl
Socorex/Wheaton Calibra/822 2-20 μl
Transferpette 2-20 μl
Transferpette Multi-Channel 5-50 μl
General description
Perfect for low-volume gel loading
Precision-molded tips with the patented Art self-sealing barrier. Guaranteed to prevent carryover contamination during aspiration and delivery of samples. Art tips are ideal for use in genetic studies, forensics, general PCR work and radioisotope sampling. Art tips are available in numerous sizes and styles to meet all your liquid handling needs. All Art tips are certified RNase-, DNase-, DNA- and pyrogen-free.
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.