Compatibility
Compatibility
Biohit mLINE 10-100 μl
Biohit Electronic Multi-Channel 5-100 μl
Biohit Multi-Channel 5-50 μl
Eppendorf Research Pro 4860 5-100 μl
Eppendorf Titerman 4908 30-300 μl
Finnpipette 4027/4500 Digital 5-40 μl
Finnpipette Multi-Channel 5-50 μl
Hamilton SoftGrip Precision 100 μl
Nichiryo BenchMate 5-50 μl
Nichiryo BenchMate Multi-Channel 5-50 μl
Socorex/Wheaton Calibra/822 10-100 μl
General description
Molded with reference marks at 10, 50, and 100 μl
Precision-molded tips with the patented Art self-sealing barrier. Guaranteed to prevent carryover contamination during aspiration and delivery of samples. Art tips are ideal for use in genetic studies, forensics, general PCR work and radioisotope sampling. Art tips are available in numerous sizes and styles to meet all your liquid handling needs. All Art tips are certified RNase-, DNase-, DNA- and pyrogen-free.
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.