Compatibility
Compatibility
Biohit Electronic 10-250 μl
Biohit Electronic Multi-Channel 25-250 μl
Biohit Variable 50-250 μl
Eppendorf Response 4850 20-500 μl
Eppendorf Research Pro 4860 20-300 μl
Eppendorf Titerman 4908 30-300 μl
Finnpipette Multi-Channel 50-300 μl
Hamilton SoftGrip Precision 300 μl
Rainin EDP 2 20-250 μl
Rainin EDP Plus 20-250 μl
Socorex/Wheaton Calibra/822 20-200 μl
Transferpette Multi-Channel 20-200ul/30-300 μl
General description
Precision-molded tips with the patented Art self-sealing barrier. Guaranteed to prevent carryover contamination during aspiration and delivery of samples. Art tips are ideal for use in genetic studies, forensics, general PCR work and radioisotope sampling. Art tips are available in numerous sizes and styles to meet all your liquid handling needs. All Art tips are certified RNase-, DNase-, DNA- and pyrogen-free.
A secure fit and extra space for repeat pipetting
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.