Compatibility
Compatibility
Eppendorf 4810 50-250 μl
Biohit Electronic 50-500 μl
Transferpette 25-250 μl
General description
SoftFit¿ design creates a secure fit that is easier to load than normal tips
Precision-molded tips with the patented Art self-sealing barrier. Guaranteed to prevent carryover contamination during aspiration and delivery of samples. Art tips are ideal for use in genetic studies, forensics, general PCR work and radioisotope sampling. Art tips are available in numerous sizes and styles to meet all your liquid handling needs. All Art tips are certified RNase-, DNase-, DNA- and pyrogen-free.
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.