General description
Includes 2 large and 4 small gel trays, 6 polyacrylamide gel covers, casting plate for 6 gels and 2 dual format combs
The Gel XL-100™ mini-gel electrophoresis system is designed for low voltage separation of nucleic acids and proteins in both agarose and polyacrylamide gel matrices. All components for quickly casting and running horizontal gels are supplied with the system including a gel casting set, gel box and direct-connect power supply.
• Ideal for separation of nucleic acids and proteins
• Cast and run agarose and acrylamide gels in the same apparatus
• Save time by pouring up to 6 mini-gels at once without tape or spacers
• Leakproof gel box with interlocking safety lid provides maximum safety
Specifications:
Gel Casting Set
Capacity 2 large and 4 small gel trays
Dimensions 126 × 240 × 28 mm (L × W × H)
Power Supply
Input 120 VAC, 50/60 Hz
Output 50 or 100 VDC, selectable
Fuse 0.8A
Dimensions 104 × 40 × 60 mm (L × W × H)
Migration Tank
Gel Capacity 1 large (107 × 26 mm)
2 small (52 × 60 mm)
Safety Mechanical lid interlock
Dimensions 137 × 199 × 75 mm (L × W × H)
Legal Information
Gel XL-100 is a trademark of Labnet Intl., Inc.