General description
Easy to set up and safe to use, the Gel XL Ultra V-2 includes all components necessary for casting and running agarose gels. The unit is compact, requiring a minimal amount of bench space. The direct-connect power supply eliminates a tangled mess of leads and cords on the bench and saves space. Designed for safety and convenience, the power supply can detect when the gel box cover is absent or removed, and cuts power to the system. Included in the Gel XL Ultra V-2 system is the direct-connect, intelligent power supply, UV transparent gel tank with safety lid, one large gel tray (13 × 12 cm), two small gel trays (13 × 6 cm), 4 dual format combs (13/26 teeth) and a standard casting stand. The self-sealing casting stand holds one large or two small gel trays and offers the most convenient method of gel casting available.
The Gel XL Ultra V-2 system's unique power supply connects directly on to the side of the gel tank ensuring a proper connection every time. The power supply is easily removed for thorough cleaning of the gel tank. Seven output voltages can be chosen to optimize separation conditions. A digital timer controls operation, shutting down output voltage and sounding an alarm at the expiration of time. The last used voltage and timer settings are held in memory for repetitive runs. A lid interlock located on the side of the power supply detects the presence or absence of the safety cover. If the cover is not in place or removed during a run, the power supply will not operate. Lid interlock shuts off power when lid is removed The unit carries the CE mark.
Legal Information
Gel XL Ultra V-2 is a trademark of Labnet Intl., Inc.