General description
Power Station 300 Plus is the same as the Power Station 300, but with an extended current range, from 4 to 500mA.
Power Station power supplies are ideal for DNA, RNA and protein electrophoresis as well as blotting. They provide a high degree of sophistication and versatility yet are extremely easy to program. Three models are available to meet a variety of user needs.
• Intelligent control: The sophisticated microprocessor in the Power Station power supplies utilizes feedback loops to maintain tight control over operating parameters. A number of safety features are incorporated into the software including automatic crossover, no-load detection, sudden load change and over voltage protection. Operation of the power supplies can be timed or continuous, for running up to four gels at one time.
• Constant voltage or constant current: Power Station power supply output can be set to either constant voltage or constant current with automatic crossover when the preset limit for either parameter is reached. In the event of a power failure, the unit can be programmed to resume the run with the original settings upon restoration of power. An error code will be shown in the display to alert the user that a power failure has occurred.
• Easy to use: Power Station power supplies are easy to use with very straight forward and simple programming. All operating parameters, including voltage, current and time, are set using the membrane keypad and displayed digitally. The large display is visible from across the laboratory. Four sets of output terminals allow for the simultaneous running of multiple gels from the same power supply. A retractable stand on the bottom allows the power supply to be placed at an angle for easier viewing and access.
Legal Information
Power Station is a trademark of Labnet Intl., Inc.