Preface
Chapter 1: The Origins, Evolution, and Directions of Human Gene Therapy (T. Friedmann)
Chapter 2: Retroviral Vectors (J.-K. Yee)
Chapter 3: Lentiviral Vectors (L. Naldini and I.M. Verma)
Chapter 4: Structure and Genetic Organization of Adenovirus Vectors (M.M Hitt, R.J. Parks, and F.L. Graham)
Chapter 5: Adenovirus Vectors (N.A. Wivel, G.-P. Gao, and J.M. Wilson)
Chapter 6: Strategies to Adapt Adenoviral Vectors for Gene Therapy Applications: Targeting and Integration (P.N. Reynolds and D.T. Curiel)
Chapter 7: Adeno-associated Viral Vectors (R.J. Samulski, M. Sally, and N. Muzyczka)
Chapter 8: Gene-transfer Tool: Herpes Simplex Virus Vectors (S. Laquerre, W.F. Goins, S. Moriuchi, T.J. Oligino, D.M. Krisky, P. Marconi, M.K Soares, J.B. Cohen, J.C. Glorioso, and D.J. Fink)
Chapter 9: Emerging Viral Vectors (D.J. Jolly)
Chapter 10: Advances in Synthetic Gene-delivery System Technology (P.L. Felgner, O. Zelphati, and X. Liang)
Chapter 11: Receptor-mediated Gene Delivery Strategies (M. Cotten and E. Wagner)
Chapter 12: Naked DNA Gene Transfer in Mammalian Cells (J.A. Wolff)
Chapter 13: Delivery Systems and Adjuvants for DNA Vaccines (J.B. Ulmer and M.A. Liu)
Chapter 14: Targets for Gene Therapy (T. Friedmann and A.G. Motulsky)
Chapter 15: The Hematopoietic System as a Target for Gene Therapy (B.P. Sorrentino and A.W. Nienhuis)
Chapter 16: Cancer Immunotherapy (D. Pardoll and G.J. Nabel)
Chapter 17: Gene Transfer and Stem-cell Transplantation (M.K. Brenner)
Chapter 18: Suicide Genes: Gene Therapy Applications Using Enzyme/Prodrug Strategies (J.C. Morris, R. Touraine, O. Wildner, and R.M. Blaese)
Chapter 19: The Logic of Anti-angiogenic Gene Therapy (J. Folkman, P. Hahnfeldt, and L. Hlatky)
Chapter 20: Apoptosis as a Goal of Cancer Gene Therapy (J.C. Reed)
Chapter 21: Advances in Gene Therapy for HIV and Other Viral Infections (E.M. Poeschla and F. Wong-Staal)
Chapter 22: Progress Toward Gene Therapy for Nervous System Diseases