General description
• Economy micro centrifuge
• Ultra compact with small footprint
• Non-refrigerated
• Capacity: 24 x 1.5/2.0 mL
• Max RPM/RCF: 14,000/17,350 (Fixed Angle)
• Dimensions (HxWxD): 199 mm (8.0 in.) x 231 mm (9.0 in.) x 292 mm (11.5 in.)
• 12, 18, or 24 place exchangeable rotors
• Auto-imbalance detection and shut-off
• Motor overheating protection
• Weight: 6 kg (13.25 lbs)
• CE Compliant
Packaging
CSA or cTUVus Label for Canada (SCC)
Legal Information
Hettich is a registered trademark of Hettich GmbH.
MIKRO is a trademark of Hettich GmbH.