General description
• Medium capacity universal benchtop centrifuge
• Available in non-refrigerated (38) and refrigerated (38R) versions
• Temperature Range (38R): -20 to +40°C
• Capacity: 4 x 250 mL
• Max RPM/RCF: 15,000/24,400 (Fixed Angle)
• Specialty applications available:
-Cytology up to 8 slide carriers
-Microtiter and deep well plates
-Bottles up to 250 mL
• Certified bio-containment lids available
• CE Compliant
Packaging
CSA or cTUVus Label for Canada (SCC)
Legal Information
Hettich is a registered trademark of Hettich GmbH.
ROTINA is a trademark of Hettich GmbH.