General description
The vials are clicked into the low profile microplate format rack to allow for automatic manipulation and automated transport without the risk of loosing samples. The cap is designed with a socket to allow automated manipulation through a click of the screwdriver into the cap. Designed for low temperature storage of biological materials. Developed for automated handling. Dense storage format for optimized freezer utilization Working volume of 0.5 or 1.0 ml C. Bank-It™ for low DNA adsorption surface for storage of DNA .
Legal Information
Nunc is a registered trademark of Nalge Nunc International.