General description
The model KDS330 is designed to emulsify viscous fluids/suspensions by forcing them back and forth through a micro-emulsifying needle. The pump eliminates the fatigue and time required to manually prepare the emulsion. The KDS330 is ideal to prepare an adjuvant/antigen mixture to the correct viscosity ready for injection.
The pump is based on the KDS210 in continuous mode for cycling back and forth and is specifically designed for a 10 cc glass syringe and emulsion needle.
• Simple control using a keypad, menu selection and a LCD display
• Volume setting and automatic cycling
• Rate setting
• Settings stored in memory
• Stall detection