General description
These hermetically sealed tubes prevent the spreading of aerosols or vapours created by mechanical stirring, dispersion or grinding infectious, toxic or odourous substances in open vessels. Disposable and single-use tubes, they avoid the possibility of cross-contamination of samples and remove the need for clean up of disperser tools, stirrer tools, grinding chambers or vessels. Each tube has a bayonet fitting on the bottom, for easy, secure connection to the tube drive.
With stainless steel balls for dry and brittle samples or cell disruption.
Warning
Stirring, dispersing or grinding may generate heat or cause other changes in the sample that might lead to build up of pressure inside the tube. Use appropriate personal protective equipment when operating and opening tubes.
Legal Information
IKA is a registered trademark of IKA Works, Inc.
ULTRA-TURRAX is a registered trademark of IKA Works, Inc.