General description
• Self-contained unit, ready to plug into standard light socket.
• Eliminates need to replace bulbs or filters
• Designed for Autoradiography & Electron Microscopy films
Has cluster of 20 light emitting diodes, spectrally matched to transmit light at the lowest spectral sensitivity of autoradiography & electron micrography films, delivering illumination without the heat generated by incandescent bulbs and without the risk of film fogging from worn or damaged filters. Simply screw into existing Safelight housing with filter removed, or fit in any standard light socket in your darkroom
Legal Information
Kodak is a registered trademark of Eastman Kodak Company.