General description
This handbook compiles the most comprehensive information on over 200 water-based and solvent-based indicators. Organized alphabetically, entries include: common name, other names, CA index name, CAS registry number, Merck index number, chemical structure, chemical/dye class, molecular formula, molecular weight, pH range, color change at pH, pKa, physical form, solubility, UV-visible, melting point, and boiling point. This resource also offers unique coverage including protocols for synthesizing indicator compounds; data relating to adverse effects, toxicity, and major applications for each indicator. The handbook contains practical information for applications that include semiconductors, fuel cells, sensors, surface coatings, adhesives, agricultural chemicals, textiles, personal care products, pharmaceuticals, and detection of disease.