General description
Bransonic cleaners are used routinely to clean electronics components, contact lenses, and gemstones. You can be sure they will safely clean your glassware and system components. Recessed cleaning tank is enclosed in durable, solvent and impact resistant plastic. The capaciity is ˝ gallon (1.8 liter). Ultrasonic cleaning is fast, effective and safe. The Branson ultrasonic cleaner has more power than most comparable models. It ensures faster, more thorough cleaning of dirt, protein residue, and other debris from your glassware, fittings, syringes, needles, and other apparatus. It also is useful for ultrasonic digestion of environmental samples.
Legal Information
Bransonic is a registered trademark of Branson Ultrasonics Corp.