General description
Retention Mechanism: Normal phase
Sample Matrix Compatibility: Organic solvents, oils, and lipids
• Unbonded acid washed silica sorbent ideal for normal phase SPE and other modified flash techniques
• Often used to separate or remove structurally similar molecules through successive elutions with increasingly polar solutions
• The most polar normal phase sorbent available
• Excellent capacity for purifying solution phase combinatorial chemistry reactions when removing target molecules from reaction by-products and excess reagents
• Available in Büchner Funnel configurations for easy scalability
Legal Information
Discovery is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.