| 59592 Supelco |
SUPELCOSIL™ LC-304 Supelguard™ Cartridge5 μm particle size, L × I.D. 2 cm × 4.0 mm |
|
|||||||||
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. |