Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Product No. Only)
Supplier X Reference
Substructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
861259
Supelco
Acid Herbicide Spiking Mix 2
in acetone (varied)
Expand/Collapse All
Price and Availability
Properties
feature
standard type spiking
packaging
pkg of 10 mL
concentration
in acetone (varied)
Safety
Hazard Codes
F,Xi
Risk Statements
11-36-66-67
Safety Statements
9-16-26
RIDADR
UN 1090 3/PG 2
Components
Analyte
2,4-Dichlorophenoxyacetic acid 50 μg/mL
2-(2,4,5-Trichlorophenoxy)propionic acid
2,4,5-Trichlorophenoxyacetic acid
Related Categories
...
A
>
AA to AL
...
8000 Series Solidwaste Methods
>
Method 8150/8151
Scroll Up
Scroll Down
Related Information
MSDS
Certificate of Analysis
More Information
Links
Related Categories
Page Options
Printer Friendly View
Bulk Quote
Ask A Scientist
Last 5 Products Viewed
861259 (Supelco)
en-us-footer US
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.