Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Product No. Only)
Supplier X Reference
Substructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
861283
Supelco
OP Pesticide Mix 2
1000 μg/mL each component in acetone, pkg of 1 mL
Expand/Collapse All
Price and Availability
Properties
packaging
pkg of 1 mL
concentration
1000 μg/mL each component in acetone
suitability
suitable for 8141 per US EPA
Safety
Hazard Codes
F,Xi
Risk Statements
11-36-66-67
Safety Statements
9-16-26
RIDADR
UN 1090 3/PG 2
Components
Analyte
Alachlor
Atrazine
Bromacil
Butachlor
Metolachlor
Prometon
Simazine
Related Categories
...
Alphabetic
>
O
Scroll Up
Scroll Down
Related Information
MSDS
Certificate of Analysis
More Information
Links
Related Categories
Page Options
Printer Friendly View
Bulk Quote
Ask A Scientist
Last 5 Products Viewed
861283 (Supelco)
en-us-footer US
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.