General description
The VersaFlash® station secures the VersaPak® cartridges for processing. The unique pre-compressed cartridge design eliminates the need for compression barrels making cartridge change-out fast and easy. Installing, removing, and changing out VersaPak cartridges in the station is fast and easy by simply turning and pulling out the handle. Scale-up to larger cartridges is just as easy by simply loosening the knob on the upper platen and adjusting it to the proper height for the larger or smaller cartridge. Most flash chromatography systems typically limit users to a narrow sample range, requiring additional hardware purchase and laborious change-out in order to work with larger sample sizes. The VersaFlash station is available separately or as a complete system with a pump, VersaVac sample loading station, and starter kit.
The unique design of the VersaFlash station also makes it possible to:
• Extend cartridge length by stacking two cartridges together.
• Load low solubility samples directly in-line using a solid sample cartridge.
• Achieve bimodal separations by stacking different phase chemistries.
• Elute strongly retained substances quickly by using the Rev-Elution technique.
The VersaFlash™ station secures the VersaPak™ cartridges for processing. The unique pre-compressed cartridge design eliminates the need for compression barrels making cartridge change-out fast and easy. Installing, removing, and changing out VersaPak cartridges in the station is fast and easy by simply turning and pulling out the handle. Scale-up to larger cartridges is just as easy by simply loosening the knob on the upper platen and adjusting it to the proper height for the larger or smaller cartridge. Most flash chromatography systems typically limit users to a narrow sample range, requiring additional hardware purchase and laborious change-out in order to work with larger sample sizes. The VersaFlash station is available separately or as a complete system with a pump, VersaVac sample loading station, and starter kit.
Legal Information
VersaPak is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.
VersaFlash is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.