Features and Benefits
Adsorption area: 100cm2
General description
Ready-to-use Sartobind membrane units are the ultimate for simple, ultra-rapid concentration of proteins from highly dilute solutions, or for separating proteins from a mixture. They feature high binding capacity (~1 mg/cm3), high flow rates (>100 mL/min.), and excellent resolution. Use these units with syringes, peristaltic pumps, or preparative HPLC/FPLC workstations. the disposable 5 cm2 units are very useful for rapidly determining buffer conditions for ion exchange separations - simply connect units with different functionalities in series.
Female Luer-Lock inlet / Male Luer-Lock outlet design.
Legal Information
Sartobind is a trademark of Sartorius AG.