Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Product No. Only)
Supplier X Reference
Substructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
Supelco
Acrolein/Acrylonitrile Mix
2000 μg/mL each component in H
2
O
Expand/Collapse All
Price and Availability
Properties
description
Separate Source
feature
standard type calibration
packaging
pkg of 1 × 1 mL (46870-U)
pkg of 1 × 1 mL (4S6870-U)
concentration
2000 μg/mL each component in H
2
O
suitability
suitable for 8260 per US EPA
storage temp.
2-8°C
Components
Analyte
Acrolein
Acrylonitrile
Related Categories
...
A
>
AA to AL
...
Separate Source Standards
>
EPA 8000 Series
...
8000 Series Solidwaste Methods
>
Method 8260
...
Environmental Standards
>
Solid Waste
Scroll Up
Scroll Down
Related Information
MSDS
Certificate of Analysis
More Information
Links
Similar Products
Related Categories
Page Options
Printer Friendly View
Bulk Quote
Ask A Scientist
Last 5 Products Viewed
en-us-footer US
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.