• USA Home
  • Search Results

  • 18crown6264321745513911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

18-Crown-6

4 Products
Synonym:   1,4,7,10,13,16-Hexaoxacyclooctadecane
Image of 18-Crown-6
CAS Number:
Empirical Formula (Hill Notation):
 C12H24O6
Molecular Weight:
 264.32
 Beilstein Registry Number:
 1619616
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Bulletin (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More