• USA Home
  • Search Results

  • 1methylimidazole821061647711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Methylimidazole

4 Products
Synonym:   N-Methylimidazole
Image of 1-Methylimidazole
CAS Number:
Linear Formula:
 C₄H₆N₂
Molecular Weight:
 82.10 g/mol
 EC Number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
Show More