• USA Home
  • Search Results

  • 1r8s9sbicyclo610non4yn9ylmethanol15022126316690011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethanol

1 Products
Synonym:   (1α,8α,9β)-Bicyclo[6.1.0]non-4-yne-9-methanol, endo-9-Hydroxymethylbicyclo[6.1.0]non-4-yne, BCN-OH
Image of (1<I>R</I>,8<I>S</I>,9<I>s</I>)<WBR>-<WBR>Bicyclo[6.1.0]<WBR>non-<WBR>4-<WBR>yn-<WBR>9-<WBR>ylmethanol
CAS Number:
Empirical Formula (Hill Notation):
 C10H14O
Molecular Weight:
 150.22
 Beilstein Registry Number:
 21037713
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Chemical Composition
Show More